Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icosl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015790.3 | NP_056605.1 | 7 | 1531 | ||
GenBank mRNA | AF199027.1 | - | 7 | 1440 | ||
AK041421.1 | - | 7 | 1541 | |||
AK136074.1 | - | 4 | 714 | |||
AK220220.1 | - | 4 | 1258 | |||
BC029227.1 | - | 7 | 1531 | 4217333 |