Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015811.2 | NP_056626.2 | 4 | 402 | ||
GenBank mRNA | AF215667.1 | - | 4 | 333 | ||
AK089293.1 | - | 3 | 869 | |||
AK138688.1 | - | 4 | 358 | |||
AK154394.1 | - | 358 | ||||
AK154841.1 | - | 2758 | ||||
AK170366.1 | - | 4 | 354 | |||
AK170957.1 | - | 4 | 358 | |||
AK171306.1 | - | 699 | ||||
BC028634.1 | - | 4 | 333 | 1328269 | ||
BC052684.1 | - | 2 | 1188 |