Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025427.3 | NP_001020598.1 | 5 | 1049 | ||
NM_001039356.2 | NP_001034445.1 | 6 | 1164 | |||
NM_001166535.1 | NP_001160007.1 | 5 | 1087 | |||
NM_001166536.1 | NP_001160008.1 | 5 | 1047 | |||
NM_001166537.1 | NP_001160009.1 | 5 | 1341 | |||
NM_001166539.1 | NP_001160011.1 | 6 | 1131 | |||
NM_001166540.1 | NP_001160012.1 | 5 | 1054 | |||
NM_001166541.1 | NP_001160013.1 | 5 | 1016 | |||
NM_001166542.1 | NP_001160014.1 | 5 | 1014 | |||
NM_001166543.1 | NP_001160015.1 | 5 | 1084 | |||
NM_001166544.1 | NP_001160016.1 | 5 | 1046 | |||
NM_001166545.1 | NP_001160017.1 | 5 | 1061 | |||
NM_001166546.1 | NP_001160018.1 | 5 | 1045 | |||
NM_016660.3 | NP_057869.2 | 5 | 1374 | |||
GenBank mRNA | AK154957.1 | - | 5 | 970 | ||
AK155661.1 | - | 5 | 968 | |||
AK160036.1 | - | 5 | 938 | |||
AK169783.1 | - | 5 | 962 | |||
AK171334.1 | - | 5 | 1003 | |||
AK191014.1 | - | 1 | 84 | |||
AK195902.1 | - | 1 | 84 | |||
J04179.1 | - | 5 | 1038 |
Gene Symbol: Hmga1-rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166476.1 | NP_001159948.1 | 1 | 970 | ||
NM_001166477.1 | NP_001159949.1 | 2 | 937 | |||
GenBank mRNA | BC008125.1 | - | 2 | 922 | ||
BC013455.1 | - | 1 | 980 | 4219805 | ||
BC093496.1 | - | 1 | 937 | 3601855 |