Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016688.2 | NP_057897.2 | 1 | 928 | ||
GenBank mRNA | AF083929.1 | - | 1 | 922 | ||
AK155050.1 | - | 1 | 893 | |||
BU518553.1 | - | 1 | 342 |