Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Incenp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016692.3 | NP_057901.2 | 2 | 301 | ||
GenBank mRNA | AB100432.1 | - | 1 | 95 | ||
AB100433.1 | - | 1 | 95 | |||
AF117610.1 | - | 2 | 237 | |||
AK034764.1 | - | 2 | 230 | |||
AK081841.1 | - | 2 | 235 | |||
AK088627.1 | - | 2 | 258 | |||
BC021761.2 | - | 2 | 198 | |||
BC037011.1 | - | 2 | 254 | |||
BC052414.1 | - | 2 | 234 | 6401188 |