Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uchl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016723.2 | NP_057932.2 | 3 | 219 | ||
GenBank mRNA | AB033370.1 | - | 3 | 192 | ||
AF247358.1 | - | 129 | ||||
AK050278.1 | - | 3 | 219 | |||
BC029242.1 | - | 3 | 161 | |||
BC048481.1 | - | 3 | 176 | 6773555 |