Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164156.1 | NP_001157628.1 | 2, 3 | 247 | ||
NM_016766.3 | NP_058046.2 | 3, 4 | 367 | |||
GenBank mRNA | AF015309.1 | - | 3, 4 | 317 | ||
AK088116.1 | - | 3, 4 | 332 | |||
AK158347.1 | - | 3, 4 | 347 | |||
AK167242.1 | - | 2, 3 | 207 | |||
BC003746.1 | - | 3, 4 | 351 | 3486681 | ||
BC085099.1 | - | 3, 4 | 335 | 6391692 | ||
BC108340.1 | - | 3, 4 | 342 | 4504579 | ||
CN724575.1 | - | 3, 4 | 367 |