Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016776.2 | NP_058056.2 | 1, 2 | 199 | ||
GenBank mRNA | AK083908.1 | - | 1, 2 | 223 | ||
AK143487.1 | - | 1, 2 | 232 | |||
AK154016.1 | - | 1, 2 | 221 | |||
AK155074.1 | - | 1, 2 | 210 | |||
AK166533.1 | - | 1, 2 | 223 | |||
AK208884.1 | - | 1, 2 | 103 | |||
BC015070.1 | - | 1, 2 | 200 | |||
BC048858.1 | - | 1, 2 | 199 | 5363740 | ||
BC052889.1 | - | 1, 2 | 220 | 30057169 | ||
BC060167.1 | - | 1, 2 | 205 | |||
U63648.1 | - | 1, 2 | 208 |