Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crlf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164735.1 | NP_001158207.1 | 8 | 1057 | ||
NM_016715.4 | NP_057924.3 | 8 | 1024 | |||
GenBank mRNA | AB031333.1 | - | 8 | 972 | ||
AB039945.1 | - | 8 | 920 | |||
AF201963.1 | - | 8 | 1108 | |||
AF232936.1 | - | 8 | 988 | |||
AK010291.1 | - | 3 | 216 | |||
AK149928.1 | - | 8 | 1026 | |||
AK170466.1 | - | 5 | 886 | |||
BC016580.1 | - | 8 | 1102 | 2655857 | ||
BC023788.1 | - | 8 | 1012 | 5346748 |