Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016737.2 | NP_058017.1 | 2 | 300 | ||
GenBank mRNA | AK075988.1 | - | 2 | 238 | ||
AK088494.1 | - | 2 | 242 | |||
AK134171.1 | - | 2 | 216 | |||
AK149493.1 | - | 2 | 216 | |||
AK161645.1 | - | 2 | 219 | |||
AK167273.1 | - | 2 | 234 | |||
AK168181.1 | - | 2 | 216 | |||
BC003794.1 | - | 2 | 209 | 3590457 | ||
CJ155828.1 | - | 2 | 301 | |||
U27830.1 | - | 2 | 217 |