Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122982.2 | NP_001116454.1 | 2 | 296 | ||
NM_001290420.1 | NP_001277349.1 | 2 | 296 | |||
NM_001290422.1 | NP_001277351.1 | 2 | 296 | |||
NM_016746.4 | NP_058026.2 | 2 | 296 | |||
GenBank mRNA | AK009615.1 | - | 2 | 241 | ||
AK014079.1 | - | 2 | 230 | |||
AK035728.1 | - | 2 | 241 | |||
AK035762.1 | - | 2 | 257 | |||
AK037957.1 | - | 2 | 297 | |||
AK135472.1 | - | 2 | 292 | |||
AK153035.1 | - | 2 | 229 | |||
BB631264.1 | - | 2 | 294 | |||
BC003344.1 | - | 2 | 229 | 3257730 | ||
BC029211.1 | - | 2 | 242 | 4526925 | ||
BC037689.1 | - | 2 | 224 | |||
BC062376.1 | - | 2 | 211 | |||
BC087544.1 | - | 2 | 228 | |||
BC100396.1 | - | 2 | 270 | |||
BC120649.1 | - | 2 | 223 | 40129572 | ||
BC120677.1 | - | 2 | 223 | 40129600 | ||
U62638.1 | - | 2 | 271 |