Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243762.1 | NP_001230691.1 | 12 | 2438 | ||
NM_016691.4 | NP_057900.3 | 10 | 2056 | |||
GenBank mRNA | AF134117.1 | - | 10 | 2056 | ||
AK142227.1 | - | 10 | 1988 | |||
AK145763.1 | - | 10 | 2025 | |||
BC036347.1 | - | 10 | 2028 | 3989812 |