Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif21a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109040.2 | NP_001102510.1 | 37 | 5258 | ||
NM_001109041.2 | NP_001102511.1 | 36 | 5126 | |||
NM_001109042.2 | NP_001102512.1 | 33 | 4943 | |||
NM_016705.4 | NP_057914.2 | 33 | 4961 | |||
GenBank mRNA | AF202892.1 | - | 33 | 4821 | ||
AK047208.1 | - | 1 | 55 | |||
AK049303.1 | - | 11 | 1459 | |||
AK052821.1 | - | 1 | 14 | |||
AK053373.1 | - | 2 | 199 | |||
AK129426.1 | - | 30 | 3866 | |||
AK147436.1 | - | 36 | 5014 | |||
BC060698.1 | - | 33 | 4810 | 6409819 | ||
BC062896.1 | - | 33 | 4810 | 6409819 |