Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159289.1 | NP_001152761.1 | 2 | 667 | ||
NM_001159290.1 | NP_001152762.1 | 2 | 322 | |||
NM_001242934.1 | NP_001229863.1 | 2 | 667 | |||
NM_016707.3 | NP_057916.1 | 2 | 667 | |||
GenBank mRNA | AA008362.1 | - | 1 | 121 | ||
AF051525.1 | - | 2 | 605 | |||
AF169037.1 | - | 2 | 605 | |||
AF186018.1 | - | 2 | 548 | |||
AK004395.1 | - | 2 | 543 | |||
AK043677.1 | - | 2 | 466 | |||
AK045556.1 | - | 2 | 323 | |||
AK122557.1 | - | 2 | 576 | |||
AK140949.1 | - | 2 | 612 | |||
AK166352.1 | - | 2 | 668 | |||
BC051418.1 | - | 2 | 455 |