Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neu3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016720.2 | NP_057929.1 | 3 | 2605 | ||
GenBank mRNA | AB026842.1 | - | 3 | 2604 | ||
AK018476.1 | - | 1 | 176 | |||
AK050428.1 | - | 3 | 2483 | |||
AK134405.1 | - | 3 | 2563 |