Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF534114.1 | - | 9 | 1013 | ||
AK018166.1 | - | 9 | 1822 | |||
AK086301.1 | - | 10 | 1223 | |||
AK134581.1 | - | 10 | 1067 | |||
AK153400.1 | - | 10 | 1051 | |||
BC005640.1 | - | 10 | 1052 | |||
BC018178.1 | - | 10 | 1038 | |||
BC037451.1 | - | 9 | 1042 | 4018648 | ||
U27462.1 | - | 10 | 1260 |