Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177778.2 | NP_001171249.1 | 20 | 3820 | ||
NM_001177779.1 | NP_001171250.1 | 14 | 2817 | |||
NM_001177780.1 | NP_001171251.1 | 12 | 2209 | |||
NM_001290402.1 | NP_001277331.1 | 19 | 3766 | |||
NM_016747.4 | NP_058027.1 | 21 | 3862 | |||
GenBank mRNA | AK122474.1 | - | 20 | 3819 | ||
AK145815.1 | - | 14 | 2572 | |||
AK148464.1 | - | 12 | 2189 | |||
AK159655.1 | - | 12 | 2197 | |||
AK167365.1 | - | 12 | 2196 | |||
BC060713.1 | - | 19 | 3894 | |||
BC094368.1 | - | 19 | 3761 | 6842105 | ||
D87117.1 | - | 21 | 3836 |