Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016754.5 | NP_058034.1 | 7 | 681 | ||
GenBank mRNA | AK003149.1 | - | 7 | 586 | ||
AK010483.1 | - | 7 | 682 | |||
BC055869.1 | - | 7 | 565 | 4018694 | ||
U77943.1 | - | 6 | 528 |