Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016756.4 | NP_058036.1 | 7 | 1761 | ||
NM_183417.3 | NP_904326.1 | 8 | 1905 | |||
GenBank mRNA | AK146845.1 | - | 8 | 1885 | ||
AK151979.1 | - | 8 | 1855 | |||
AK152922.1 | - | 7 | 1714 | |||
AK153983.1 | - | 8 | 1857 | |||
AK214050.1 | - | 1 | 46 | |||
BC005654.1 | - | 8 | 1866 | 3709730 | ||
BC026145.1 | - | 6 | 2467 | |||
U63337.1 | - | 8 | 1188 |