Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164156.1 | NP_001157628.1 | 12, 13 | 1272 | ||
NM_016766.3 | NP_058046.2 | 13, 14 | 1392 | |||
GenBank mRNA | AF015309.1 | - | 13, 14 | 1342 | ||
AK088116.1 | - | 13, 14 | 1357 | |||
AK158347.1 | - | 13, 14 | 1372 | |||
AK158611.1 | - | 9, 10 | 999 | |||
AK167242.1 | - | 12, 13 | 1232 | |||
BC003746.1 | - | 13, 14 | 1376 | 3486681 | ||
BC085099.1 | - | 13, 14 | 1360 | 6391692 | ||
BC108340.1 | - | 13, 14 | 1367 | 4504579 |