Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290576.1 | NP_001277505.1 | 3, 4 | 649 | ||
NM_016768.2 | NP_058048.1 | 3, 4 | 649 | |||
GenBank mRNA | AF020199.1 | - | 3, 4 | 640 | ||
AF020200.1 | - | 3, 4 | 642 | |||
AK144363.1 | - | 3, 4 | 566 | |||
BC057879.1 | - | 3, 4 | 540 | 5348692 |