Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016769.4 | NP_058049.3 | 9 | 1757 | ||
GenBank mRNA | AF016189.1 | - | 9 | 1733 | ||
AK041743.1 | - | 9 | 1278 | |||
AK048626.1 | - | 9 | 1757 | |||
AK083158.1 | - | 9 | 1365 | |||
AK210600.1 | - | 1 | 127 | |||
BC066850.1 | - | 9 | 1443 | 30432720 |