Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110147.1 | NP_001103617.1 | 12 | 2412 | ||
NM_001289443.1 | NP_001276372.1 | 4 | 799 | |||
NM_016788.3 | NP_058068.2 | 13 | 2457 | |||
GenBank mRNA | AF037260.1 | - | 13 | 2240 | ||
AK087965.1 | - | 9 | 1569 | |||
BC031168.1 | - | 4 | 806 | 4984168 | ||
BC052421.1 | - | 12 | 2412 | 6400209 | ||
DQ666696.1 | - | 13 | 2268 |