Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016803.3 | NP_058083.2 | 3 | 688 | ||
GenBank mRNA | AB008937.1 | - | 2 | 307 | ||
BC055055.1 | - | 1 | 567 | 6493234 |