Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdcbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016807.2 | NP_058087.2 | 4, 5 | 344 | ||
GenBank mRNA | AF077527.1 | - | 4, 5 | 303 | ||
AK150661.1 | - | 4, 5 | 312 | |||
AK150736.1 | - | 4, 5 | 312 | |||
AK159323.1 | - | 4, 5 | 290 | |||
AK159538.1 | - | 4, 5 | 317 | |||
AK165657.1 | - | 4, 5 | 314 |