Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005729.3 | NP_005720.1 | 2 | 294 | ||
XM_005269379.2 | XP_005269436.1 | 2 | 292 | |||
GenBank mRNA | AK296669.1 | - | 2 | 287 | ||
BC005020.1 | - | 2 | 280 | 3659599 | ||
BC013204.1 | - | 2 | 271 | |||
BC110299.1 | - | 2 | 271 | 5215228 | ||
BE791220.1 | - | 2 | 310 | |||
M80254.1 | - | 2 | 292 |