Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acrbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127340.1 | NP_001120812.1 | 4 | 496 | ||
NM_016845.2 | NP_058541.2 | 4 | 496 | |||
GenBank mRNA | AK076691.1 | - | 4 | 461 | ||
AK132705.1 | - | 4 | 464 | |||
BC011079.1 | - | 4 | 734 | 3984550 | ||
BC114591.1 | - | 4 | 465 | |||
BM938496.1 | - | 3 | 269 | |||
D17573.1 | - | 4 | 448 | |||
D17574.1 | - | 4 | 500 |