Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016846.3 | NP_058542.2 | 2 | 173 | ||
GenBank mRNA | AK144218.1 | - | 2 | 248 | ||
AK169109.1 | - | 2 | 225 | |||
AK169180.1 | - | 2 | 226 | |||
BC017678.1 | - | 2 | 173 | 4241244 | ||
U14103.1 | - | 2 | 231 |