Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016852.2 | NP_058548.1 | 2 | 189 | ||
GenBank mRNA | AK004824.1 | - | 2 | 191 | ||
AK043037.1 | - | 2 | 163 | |||
AK079300.1 | - | 2 | 179 | |||
AK146572.1 | - | 2 | 191 | |||
AK150018.1 | - | 2 | 164 | |||
AK151368.1 | - | 2 | 181 | |||
AK162810.1 | - | 2 | 164 | |||
AK170317.1 | - | 2 | 190 | |||
AK170474.1 | - | 2 | 192 | |||
AK187444.1 | - | 2 | 119 | |||
BC031908.1 | - | 2 | 152 | 5365118 | ||
BC055058.1 | - | 2 | 209 | 6529458 | ||
U40826.1 | - | 2 | 189 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK044015.1 | - | 179 | Chr. - - [-] on Build |