Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297644.1 | NP_001284573.1 | 6 | 767 | ||
NM_001297645.1 | NP_001284574.1 | 6 | 767 | |||
NM_001318000.1 | NP_001304929.1 | 6 | 767 | |||
NM_005869.3 | NP_005860.2 | 6 | 767 | |||
XM_011543095.2 | XP_011541397.2 | 6 | 736 | |||
XM_011543096.1 | XP_011541398.1 | 6 | 736 | |||
XM_017008943.1 | XP_016864432.1 | 6 | 736 | |||
GenBank mRNA | AF039692.1 | - | 6 | 719 | ||
AF039693.1 | - | 2 | 158 | |||
AK225105.1 | - | 6 | 734 | |||
AK225161.1 | - | 6 | 739 | |||
AK308223.1 | - | 6 | 736 | |||
AK312954.1 | - | 6 | 736 | |||
AY358569.1 | - | 6 | 728 | |||
BC012117.1 | - | 6 | 720 | 4552442 | ||
BX647971.1 | - | 6 | 768 |