Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005813.4 | NP_005804.1 | 16 | 2897 | ||
XM_005264237.3 | XP_005264294.1 | 17 | 3170 | |||
XM_005264238.3 | XP_005264295.1 | 16 | 2146 | |||
XM_017003741.1 | XP_016859230.1 | 18 | 3175 | |||
XM_017003742.1 | XP_016859231.1 | 17 | 2214 | |||
XM_017003743.1 | XP_016859232.1 | 14 | 1896 | |||
XM_017003744.1 | XP_016859233.1 | 13 | 1644 | |||
GenBank mRNA | AB015982.2 | - | 16 | 2897 | ||
AK027585.1 | - | 13 | 1706 | |||
AK122883.1 | - | 9 | 2571 |