Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193619.1 | NP_001180548.1 | 3 | 537 | ||
NM_016887.6 | NP_058583.1 | 2 | 859 | |||
GenBank mRNA | AF087825.1 | - | 2 | 353 | ||
AK002924.1 | - | 2 | 780 | |||
AK010558.1 | - | 2 | 779 | |||
AK087296.1 | - | 2 | 777 | |||
AK145504.1 | - | 2 | 860 | |||
AK167250.1 | - | 2 | 780 | |||
BC008104.1 | - | 2 | 424 | 3157774 | ||
BC050007.1 | - | 2 | 772 | 6530942 |