Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252614.1 | NP_001239543.1 | 6 | 853 | ||
NM_016893.5 | NP_058589.2 | 5 | 764 | |||
GenBank mRNA | AB025198.1 | - | 2 | 453 | ||
AK048520.1 | - | 5 | 902 | |||
AK051811.1 | - | 5 | 721 | |||
AK134537.1 | - | 5 | 857 | |||
AK163494.1 | - | 5 | 880 | |||
AK184785.1 | - | 2 | 246 | |||
BC010666.1 | - | 6 | 832 | 3594582 |