Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec61a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016906.4 | NP_058602.1 | 3 | 203 | ||
GenBank mRNA | AB032902.1 | - | 3 | 215 | ||
AF145253.1 | - | 3 | 125 | |||
AF222743.1 | - | 3 | 125 | |||
AK088473.1 | - | 3 | 204 | |||
AK153055.1 | - | 3 | 193 | |||
AK159349.1 | - | 3 | 191 | |||
AK167484.1 | - | 3 | 133 | |||
AK219834.1 | - | 2 | 46 | |||
BC003707.1 | - | 3 | 129 | 3592772 |