Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blcap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016916.3 | NP_058612.3 | 2 | 606 | ||
GenBank mRNA | AB037891.1 | - | 2 | 561 | ||
AK011673.1 | - | 2 | 583 | |||
AK018127.1 | - | 2 | 636 | |||
AK170338.1 | - | 2 | 583 | |||
BC026935.1 | - | 2 | 570 | 3597231 | ||
BC083339.1 | - | 2 | 570 | 6832329 | ||
BM950751.1 | - | 2 | 547 | |||
BY718283.1 | - | 2 | 636 |