Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col5a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016919.2 | NP_058615.1 | 4, 5 | 670 | ||
GenBank mRNA | AB040491.1 | - | 4, 5 | 674 | ||
AF176645.1 | - | 4, 5 | 670 | |||
BC024367.1 | - | 4, 5 | 619 | |||
BC138417.1 | - | 4, 5 | 635 | 8861437 | ||
BC138419.1 | - | 4, 5 | 635 | 8861439 | ||
BC145269.1 | - | 4, 5 | 632 |