Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fanca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016925.3 | NP_058621.2 | 2 | 147 | ||
GenBank mRNA | AF208116.1 | - | 2 | 149 | ||
AF208118.1 | - | 2 | 149 | |||
AF208122.1 | - | 2 | 133 | |||
AF208123.1 | - | 2 | 114 | |||
AK082847.1 | - | 2 | 132 | |||
AK084990.1 | - | 2 | 132 | |||
AK166112.1 | - | 2 | 132 | |||
BC150705.1 | - | 2 | 145 | 9087616 | ||
CF537383.1 | - | 2 | 64 |