Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016805.2 | NP_058085.2 | 1, 2 | 840 | ||
GenBank mRNA | AF073992.1 | - | 1, 2 | 777 | ||
AK029944.1 | - | 1, 2 | 786 | |||
AK037517.1 | - | 1, 2 | 782 | |||
AK089058.1 | - | 1, 2 | 841 | |||
AK145499.1 | - | 1, 2 | 843 | |||
AK145682.1 | - | 1, 2 | 842 | |||
AK159084.1 | - | 1, 2 | 809 | |||
AK159614.1 | - | 1, 2 | 845 | |||
AK159956.1 | - | 1, 2 | 782 | |||
AK162264.1 | - | 1, 2 | 845 | |||
AK165844.1 | - | 1, 2 | 840 | |||
AK167200.1 | - | 1, 2 | 842 | |||
AK168657.1 | - | 1, 2 | 782 | |||
BC018353.1 | - | 1, 2 | 685 | 3584692 |