Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166409.2 | NP_001159881.1 | 3 | 383 | ||
NM_001166410.2 | NP_001159882.1 | 3 | 383 | |||
NM_001166411.2 | NP_001159883.1 | 3 | 383 | |||
NM_001293658.1 | NP_001280587.1 | 3 | 383 | |||
NM_016809.6 | NP_058089.2 | 3 | 383 | |||
GenBank mRNA | AB016424.1 | - | 3 | 210 | ||
AK003720.1 | - | 3 | 201 | |||
AK007354.1 | - | 3 | 201 | |||
AK049575.1 | - | 3 | 201 | |||
AK088031.1 | - | 3 | 199 | |||
AK133795.1 | - | 3 | 203 | |||
AK154044.1 | - | 3 | 205 | |||
AK167800.1 | - | 3 | 199 | |||
AK167967.1 | - | 3 | 203 | |||
AK168285.1 | - | 3 | 204 | |||
AK168338.1 | - | 3 | 203 | |||
BC006580.1 | - | 3 | 178 | 3586577 | ||
BC059098.1 | - | 3 | 189 | 4013810 | ||
BC086491.1 | - | 3 | 194 | 30434553 | ||
BC087737.1 | - | 2 | 149 | |||
BC106176.1 | - | 3 | 239 | 30146829 | ||
CF912757.1 | - | 3 | 383 | |||
CN696660.1 | - | 3 | 216 | |||
DV045913.1 | - | 3 | 218 |