Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016811.2 | NP_058091.2 | 3 | 437 | ||
GenBank mRNA | AF085219.1 | - | 2 | 123 | ||
BC006713.1 | - | 3 | 437 | 3584592 | ||
JN832677.1 | - | 4 | 469 | |||
JN832678.1 | - | 3 | 456 | |||
JN832679.1 | - | 3 | 456 |