Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htatip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146049.1 | NP_001139521.1 | 2 | 445 | ||
NM_001146050.1 | NP_001139522.1 | 2 | 279 | |||
NM_001146052.1 | NP_001139524.1 | 1 | 269 | |||
NM_001146053.1 | NP_001139525.1 | 2 | 223 | |||
NM_016865.3 | NP_058561.2 | 2 | 389 | |||
GenBank mRNA | AF061972.1 | - | 1 | 373 | ||
AK008630.1 | - | 1 | 264 | |||
AK015389.1 | - | 2 | 270 | |||
AK144837.1 | - | 2 | 423 | |||
AK151254.1 | - | 2 | 310 | |||
AK152418.1 | - | 2 | 310 | |||
BB575839.2 | - | 2 | 223 | |||
BC004083.1 | - | 2 | 342 | 3593077 | ||
BC017372.1 | - | 1 | 235 | 4013057 | ||
CB318388.1 | - | 1 | 272 | |||
CB593552.1 | - | 2 | 271 |