Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281929.1 | NP_001268858.1 | 4 | 826 | ||
NM_001290451.1 | NP_001277380.1 | 4 | 598 | |||
NM_016886.4 | NP_058582.3 | 4 | 826 | |||
GenBank mRNA | AB022342.1 | - | 4 | 616 | ||
AB079072.1 | - | 4 | 562 | |||
AB102775.1 | - | 4 | 562 | |||
AB102776.1 | - | 4 | 562 | |||
AF483494.1 | - | 4 | 562 | |||
AF483495.1 | - | 4 | 562 | |||
AK135043.1 | - | 4 | 579 | |||
AK138615.1 | - | 4 | 731 | |||
AK157815.1 | - | 5 | 1003 | |||
AK161163.1 | - | 4 | 646 | |||
AK220530.1 | - | 4 | 613 | |||
BC076584.1 | - | 4 | 691 | |||
BC086678.1 | - | 4 | 771 | 30668476 | ||
BC129855.1 | - | 4 | 616 | 40109751 | ||
BC129856.1 | - | 4 | 616 | 40109752 |