Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016891.3 | NP_058587.1 | 5 | 642 | ||
GenBank mRNA | AB021743.1 | - | 5 | 590 | ||
AK054239.1 | - | 5 | 602 | |||
AK078135.1 | - | 5 | 621 | |||
AK088776.1 | - | 5 | 614 | |||
AK190426.1 | - | 2 | 161 | |||
BC006606.1 | - | 5 | 642 | 3157293 | ||
BC052678.1 | - | 5 | 638 | 30062580 |