Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016892.3 | NP_058588.1 | 3 | 311 | ||
GenBank mRNA | AF121906.1 | - | 3 | 224 | ||
AF173379.1 | - | 3 | 284 | |||
AK050072.1 | - | 3 | 260 | |||
BC026938.1 | - | 3 | 278 | 4024140 | ||
BY073478.1 | - | 3 | 311 |