Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042543.2 | NP_001036008.1 | 2 | 599 | ||
NM_006529.3 | NP_006520.2 | 2 | 599 | |||
XM_011532267.2 | XP_011530569.1 | 2 | 852 | |||
XM_017008630.1 | XP_016864119.1 | 3 | 623 | |||
GenBank mRNA | BC036086.1 | - | 2 | 598 | 5286731 | |
BI600831.1 | - | 615 | ||||
U93917.1 | - | 2 | 515 |