Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016907.3 | NP_058603.2 | 2 | 594 | ||
GenBank mRNA | AF099018.1 | - | 2 | 549 | ||
AK017342.1 | - | 1 | 476 | |||
BC005769.1 | - | 1 | 481 | 2650110 |