Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159711.1 | NP_001153183.1 | 3, 4 | 334 | ||
NM_016923.2 | NP_058619.1 | 3, 4 | 388 | |||
GenBank mRNA | AB018550.1 | - | 3, 4 | 388 | ||
AF168120.1 | - | 3, 4 | 358 | |||
AK019283.1 | - | 3, 4 | 412 | |||
AY641431.1 | - | 3, 4 | 282 | |||
BC116785.1 | - | 3, 4 | 384 | 40126104 |