Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016797.4 | NP_058077.2 | 10 | 1187 | ||
GenBank mRNA | AB019212.1 | - | 10 | 857 | ||
AF056323.1 | - | 10 | 799 | |||
AK002867.1 | - | 10 | 856 | |||
AK013869.1 | - | 10 | 841 | |||
AK082927.1 | - | 10 | 827 | |||
AK142742.1 | - | 834 | ||||
AK158923.1 | - | 10 | 1188 | |||
AK166822.1 | - | 10 | 857 | |||
AK169822.1 | - | 10 | 860 | |||
AK184610.1 | - | 1 | 33 | |||
AK193627.1 | - | 1 | 33 | |||
AK196137.1 | - | 1 | 33 | |||
AK197515.1 | - | 1 | 33 | |||
AK198174.1 | - | 2 | 49 | |||
AK204540.1 | - | 2 | 54 | |||
AK209586.1 | - | 2 | 65 | |||
AK216179.1 | - | 1 | 33 | |||
AK216966.1 | - | 1 | 33 | |||
AK217289.1 | - | 1 | 33 | |||
BC132123.1 | - | 9 | 737 | 40130400 | ||
BC132125.1 | - | 9 | 737 | 40130402 |