Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016846.3 | NP_058542.2 | 18 | 2922 | ||
GenBank mRNA | AK173071.1 | - | 8 | 1517 | ||
BC017678.1 | - | 18 | 2928 | 4241244 |