Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Corin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122756.1 | NP_001116228.1 | 22 | 3248 | ||
NM_016869.3 | NP_058565.2 | 22 | 3405 | |||
GenBank mRNA | AB013874.1 | - | 22 | 3405 | ||
BC093485.1 | - | 22 | 3181 | 30648458 | ||
BC138339.1 | - | 22 | 3388 | 8861359 | ||
BC138340.1 | - | 22 | 3388 | 8861360 | ||
BC145243.1 | - | 19 | 3049 | 9053795 |